missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | 257.0°C to 264.0°C |
Color | White |
Boiling Point | 276.0°C (30.0 mmHg) |
Flash Point | 240°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Plastic Drum |
Linear Formula | C(CH2OH)4 |
Quantity | 5 kg |
Beilstein | 01,528 |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'