missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanolide, TRC

Product Code. 30428193
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428193 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428193 Supplier Toronto Research Chemicals Supplier No. TRCP22045025G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxacyclohexadecan-2-one
CAS 106-02-5
Molecular Formula C15 H28 O2
Quantity 25 g
Synonym Oxacyclohexadecan-2-one (8CI','9CI','ACI), Pentadecanoic acid','15-hydroxy-','ξ-lactone (6CI','7CI), 1,15-Pentadecanolide, 1-Oxacyclohexadecan-2-one, 15-Hydroxypentadecanoic acid lactone, 15-Pentadecanolactone, 15-Pentadecanolide, 2-Pentadecalone, cpd Supra, CPE 215, Cyclopentadecanolide, Exaltolide, Macrolide Supra, Muskalactone, NSC 36763, Pentadecalactone, Pentadecanolactone, Pentadecanolide, Pentalide, Thibetolide, ω-Pentadecalactone
InChI Formula InChI=1S/C15H28O2/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-17-15/h1-14H2
SMILES O=C1CCCCCCCCCCCCCCO1
IUPAC Name oxacyclohexadecan-2-one
Molecular Weight (g/mol) 240.38
Formula Weight 240.2089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.