missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanolide, 98%

Product Code. 11463754
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463754 100 g 100g
11453754 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463754 Supplier Thermo Scientific Alfa Aesar Supplier No. A11341.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-02-5 | C15H28O2 | 240.387 g/mol

Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-02-5
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
MDL Number MFCD00039667
InChI Key FKUPPRZPSYCDRS-UHFFFAOYSA-N
Synonym cyclopentadecanolide, exaltolide, pentadecanolide, 15-pentadecanolide, pentalide, thibetolide, pentadecan-15-olide, pentadecalactone, muskalactone, muskolactone
PubChem CID 235414
IUPAC Name oxacyclohexadecan-2-one
SMILES C1CCCCCCCOC(=O)CCCCCC1

Specifications

Melting Point 33°C to 37°C
Density 0.918
Boiling Point 137°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.47
Quantity 100 g
Beilstein 143710
Merck Index 14,3910
Solubility Information Soluble in alcohol,dipropylene glycol. Insoluble in water.
Formula Weight 240.39
Percent Purity 98%
Chemical Name or Material Pentadecanolide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.