missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanoic-d29 Acid, TRC

Product Code. 30136765
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136765 5 mg 5mg
30136847 50 mg 50mg
17988549 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30136765 Supplier Toronto Research Chemicals Supplier No. TRCP2204935MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130348-95-7
Molecular Formula C15HD29O2
Molecular Weight (g/mol) 271.58
Synonym Pentadecanoic-d29 acid (9CI), Pentadecanoic acid-d29, Perdeuteropentadecanoic acid, Pentadecanoic-D29 acid
IUPAC Name pentadecanoic-d29 acid
SMILES [2H]C(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O)([2H])C([2H])([2H])[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2
Formula Weight 271.41
Chemical Name or Material Pentadecanoic-D29 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.