missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecanoic-d29 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Pentadecanoic-D29 Acid |
| CAS | 130348-95-7 |
| Molecular Formula | C15HD29O2 |
| Quantity | 0.25 g |
| Synonym | Pentadecanoic-d29 acid (9 CI), Pentadecanoic acid-d29, Perdeuteropentadecanoic acid, Pentadecanoic-D29 acid |
| InChI Formula | InChI=1 S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2 |
| SMILES | [2 H]C(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(O)=O)([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | pentadecanoic-d29 acid |
| Molecular Weight (g/mol) | 271.58 |
| Formula Weight | 271.41 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?