missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanoic acid, 99%

Product Code. 11457636
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457636 250 g 250g
11437636 10 g 10g
11447636 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457636 Supplier Thermo Scientific Alfa Aesar Supplier No. A14664.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1002-84-2 | C15H30O2 | 242.403 g/mol

Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-84-2
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00002745
InChI Key WQEPLUUGTLDZJY-UHFFFAOYSA-N
Synonym pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure
PubChem CID 13849
ChEBI CHEBI:42504
IUPAC Name pentadecanoic acid
SMILES CCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 51°C to 54°C
Boiling Point 257°C (100 mmHg)
Flash Point 110°C (230°F)
Linear Formula CH3(CH2)13CO2H
Quantity 250 g
Beilstein 1773831
Solubility Information Soluble in ethanol.
Formula Weight 242.4
Percent Purity 99%
Chemical Name or Material Pentadecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.