missing translation for 'onlineSavingsMsg'
Learn More

Pentadecafluorooctanoic acid, 96%

Product Code. 10144240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144240 5 g Glass bottle 5g
10703181 25 g Glass bottle 25g
10391424 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10144240 Supplier Thermo Scientific Acros Supplier No. 173960050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 335-67-1 | C8HF15O2 | 414.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-67-1
Molecular Formula C8HF15O2
Molecular Weight (g/mol) 414.07
MDL Number MFCD00004174
InChI Key SNGREZUHAYWORS-UHFFFAOYSA-N
Synonym perfluorooctanoic acid, pentadecafluorooctanoic acid, perfluorocaprylic acid, pfoa, octanoic acid, pentadecafluoro, perfluoroctanoic acid, perfluoroheptanecarboxylic acid, pentadecafluoro-1-octanoic acid, pentadecafluoro-n-octanoic acid, perfluoro-n-octanoic acid
PubChem CID 9554
ChEBI CHEBI:35549
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 53.0°C to 60.0°C
Color White
Boiling Point 189.0°C to 192.0°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CF3(CF2)6CO2H
Quantity 5 g
Merck Index 15, 7272
Solubility Information Solubility in water: 3.4g/L (20°C). Other solubilities: soluble in ether and ethanol
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoic acid
Formula Weight 414.07
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Pentadecafluorooctanoic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentadecafluorooctanoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H360D-May damage the unborn child.
  • H351-Suspected of causing cancer.
  • H332-Harmful if inhaled.
  • H362-May cause harm to breast-fed children.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.