missing translation for 'onlineSavingsMsg'
Learn More

Pentachloropyridine, 98%

Product Code. 11491488
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491488 500 g 500g
11401498 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491488 Supplier Thermo Scientific Alfa Aesar Supplier No. A13842.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2176-62-7 | C5Cl5N | 251.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2176-62-7
Molecular Formula C5Cl5N
Molecular Weight (g/mol) 251.312
MDL Number MFCD00006230
InChI Key DNDPLEAVNVOOQZ-UHFFFAOYSA-N
Synonym pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
PubChem CID 16584
IUPAC Name 2,3,4,5,6-pentachloropyridine
SMILES C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 123°C to 127°C
Quantity 500 g
UN Number UN3077
Beilstein 155197
Formula Weight 251.33
Percent Purity 98%
Chemical Name or Material Pentachloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.