missing translation for 'onlineSavingsMsg'
Learn More

Pentachloropyridine, 98%, Thermo Scientific™

Product Code. 10702011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702011 100 g Glass bottle 100g
10323912 5 g Glass bottle 5g
10286110 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702011 Supplier Thermo Scientific Acros Supplier No. 160221000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2176-62-7
Molecular Formula C5Cl5N
Molecular Weight (g/mol) 251.33
MDL Number MFCD00006230
InChI Key DNDPLEAVNVOOQZ-UHFFFAOYSA-N
Synonym pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
PubChem CID 16584
IUPAC Name 2,3,4,5,6-pentachloropyridine
SMILES C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 124.0°C to 126.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5N)Cl5
Quantity 100 g
Beilstein 20, 232
Formula Weight 251.33
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Pentachloropyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Pentachloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.