missing translation for 'onlineSavingsMsg'
Learn More

Pentachloroanisole-d3 (methoxy-d3), CDN

Product Code. 30319585
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Product Code. Quantity unitSize
30319585 0.25 g 0.25g
30315701 0.1 g 0.1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30319585

Brand: CDN CDND60840.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentachloroanisole D3
CAS 1219804-52-0
Molecular Formula C7 2H3 Cl5 O
Quantity 0.25 g
Synonym Pentachloroanisole D3, Pentachloroanisole-d3, 1,2,3,4,5-Pentachloro-6-(trideuteriomethoxy)benzene, 1,2,3,4,5-Pentachloro-6-(methoxy-d3)benzene
InChI Formula InChI=1 S/C7H3Cl5O/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h1H3/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
IUPAC Name 1,2,3,4,5-pentachloro-6-(trideuteriomethoxy)benzene
Molecular Weight (g/mol) 283.382
Formula Weight 280.881 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.