missing translation for 'onlineSavingsMsg'
Learn More

Penta-2,4-dienoic Acid, TRC

Product Code. 30415981
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30415981 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30415981

Brand: Toronto Research Chemicals TRCP268363100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Pentadienoic Acid
CAS 626-99-3
Molecular Formula C5 H6 O2
Quantity 100 mg
Synonym 2,4-Pentadienoic acid, α,γ-Pentadienoic acid (3CI), 1,3-Butadiene-1-carboxylic acid, 1-Carboxy-1,3-butadiene, 1-Carboxybutadiene, Butadiene-1-carboxylic acid, NSC 16628, β-Vinylacrylic acid
InChI Formula InChI=1S/C5H6O2/c1-2-3-4-5(6)7/h2-4H,1H2,(H,6,7)
SMILES OC(=O)C=CC=C
IUPAC Name penta-2,4-dienoic acid
Molecular Weight (g/mol) 98.1
Formula Weight 98.037
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.