missing translation for 'onlineSavingsMsg'
Learn More

Pennsylvania Pesticide Mixture 334 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325680
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325680 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325680 Supplier Dr. Ehrenstorfer Supplier No. DREA50000334AL
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfenapyr∣Chlorpyrifos∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dimethomorph∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenhexamid∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264∣Myclobutanil∣Naled∣Oxamyl∣Paclobutrazol∣Parathion-methyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyridaben∣Spinetoram∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
CAS 71751-41-2, 30560-19-1, 57960-19-7, 135410-20-7, 116-06-3, 131860-33-8
Analyte or Component Names Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfenapyr∣Chlorpyrifos∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dimethomorph∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenhexamid∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264∣Myclobutanil∣Naled∣Oxamyl∣Paclobutrazol∣Parathion-methyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyridaben∣Spinetoram∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 100 μg/mL∣Acephate 100 μg/mL∣Acequinocyl 100 μg/mL∣Acetamiprid 100 μg/mL∣Aldicarb 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Captan 100 μg/mL∣Carbaryl 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Chlorfenapyr 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Clofentezine 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Daminozide 100 μg/mL∣Diazinon 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Dimethomorph 100 μg/mL∣Ethoprophos 100 μg/mL∣Etofenprox 100 μg/mL∣Etoxazole 100 μg/mL∣Fenhexamid 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Hexythiazox 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methiocarb 100 μg/mL∣Methomyl 100 μg/mL∣MGK 264 100 μg/mL∣Myclobutanil 100 μg/mL∣Naled 100 μg/mL∣Oxamyl 100 μg/mL∣Paclobutrazol 100 μg/mL∣Parathion-methyl 100 μg/mL∣Permethrin 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Propoxur 100 μg/mL∣Pyridaben 100 μg/mL∣Spinetoram 100 μg/mL∣Spinosad 100 μg/mL∣Spiromesifen 100 μg/mL∣Spirotetramat 100 μg/mL∣Spiroxamine 100 μg/mL∣Tebuconazole 100 μg/mL∣Thiacloprid 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.