missing translation for 'onlineSavingsMsg'
Learn More

Penicilline G potassium, Dr. Ehrenstorfer

Product Code. 30325120
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325120 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325120 Supplier Dr. Ehrenstorfer Supplier No. DREC15935000100MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzylpenicillin potassium
CAS 113-98-4
Analyte or Component Names Benzylpenicillin potassium
Shipping Condition +20°C
Molecular Formula C16 H17 N2 O4 S . K
Certifications/Compliance ISO 17034
Synonym Phenoxymethylpenicillin Potassium Imp. A (EP), Phenoxymethylpenicillin Imp. A (EP), Benzylpenicillin Potassium, Phenoxymethylpenicillin Potassium Imp. A (EP) as Potassium Salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid potassium salt (1:1), 3,3-Dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, [2S-(2alpha,5alpha,6beta)]-3,3-Dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, Penicillin G potassium salt, Benzylpenicillin potassium, Benzylpenicillin potassium salt, Benzylpenicillinic acid potassium salt, Cosmopen, Cristapen, Crytapen, Eskacillin, Falapen, Forpen, Hipercilina, Hyasorb, Hylenta, M-Cillin, Megacillin tablets, Monopen, NSC 131815, Notaral, Novocillin vet., Penicillin G potassium, Pentid, Pentids, Potassium 2,2-dimethyl-6beta-phenylacetamidopenam-3alpha-carboxylate, Potassium 6-(phenylacetamido)penicillanate, Potassium benzylpenicillin, Potassium benzylpenicillinate, Potassium penicillin G, Scotcil, Tabilin, BRL-4939, Benzylpenicillin potassium Pen G, K salt, Carbenicillin Sodium Impurity F as Potassium Salt, Phenoxymethylpenicillin Potassium Impurity A as Potassium Salt, Carbenicillin Impurity F as Potassium Salt, Phenoxymethylpenicillin Impurity A as Potassium Salt
InChI Formula InChI=1S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 372.48
Quantity 100 mg
Formula Weight 372.0546 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.