missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V potassium salt

Product Code. 15434819
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 15434819

Brand: Thermo Scientific Alfa Aesar J62442.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol

Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

Solubility
Soluble in water, dimethyl sulfoxide, and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-98-9
Molecular Formula C16H17KN2O5S
Molecular Weight (g/mol) 388.48
MDL Number MFCD00051771
InChI Key HCTVWSOKIJULET-LQDWTQKMSA-M
Synonym Phenoxymethylpenicillinic acid potassium salt
PubChem CID 131673927
IUPAC Name (2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin V Potassium Salt
Physical Form Powder
Melting Point 197°C to 202°C
Color White
Quantity 25 g
Beilstein 3899451
Merck Index 14,7100
Solubility Information Soluble in water,dimethyl sulfoxide,and methanol.
Formula Weight 388.48
Chemical Name or Material Penicillin V potassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.