missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Penicillin V Potassium Salt
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Inhibitor of bacterial cell wall synthesis.
Inhibitor of bacterial cell wall synthesis. Active against gram positive and some gram negative bacteria. B-Lactamase sensitive.
Specifications
Specifications
| Chemical Name or Material | Penicillin V Potassium Salt |
| CAS | 132-98-9 |
| Quantity | 100 g |
| Molecular Formula | C16H17KN2O4S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction