missing translation for 'onlineSavingsMsg'
Learn More

Penicillin/Streptomycin Mixture, Fisher BioReagents

Product Code. 10003927
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10003927 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003927 Supplier Fisher BioReagents Supplier No. BP295950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Interferes with synthesis of bacterial cell wall. Inhibits protein synthesis; binds to 30S ribosomal subunit

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Penicillin/Streptomycin Mixture
Type Penicillin/Streptomycin
Physical Form Liquid
CAS 3810-74-0, 69-57-8, 7647-14-5, 7732-18-5
Packaging Plastic Bottle
Sterile Filtered Pass Test
Quantity 50 mL
Merck Index 15, 8956
compliance-icons
Product Identifier
  • Penicillin/Streptomycin Mixture
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.