missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin Streptomycin GlutaMAX™ Supplement (100X)

Product Code. 30088326
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30088326 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30088326 Supplier Gibco™ Supplier No. A5873601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco Penicillin Streptomycin GlutaMAX Supplement is a form of GlutaMAX Supplement that contains the antibiotics penicillin (10,000 units) and streptomycin (10,000 μg), along with 200 mM L-alanyl-L-glutamine dipeptide in a citrate buffer to maintain penicillin potency.

Gibco Penicillin Streptomycin GlutaMAX Supplement is a form of GlutaMAX Supplement that contains the antibiotics penicillin (10,000 units) and streptomycin (10,000 μg), along with 200 mM L-alanyl-L-glutamine dipeptide in a citrate buffer to maintain penicillin potency. GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health.

Compared to L-glutamine, GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up
  • Improves cell viability and growth

Minimizes toxic ammonia build-up

Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth

Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

About penicillin and streptomycin

The antibiotics penicillin and streptomycin are used to prevent bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin acts by binding to the 30S subunit of the bacterial ribosome, leading to the inhibition of protein synthesis and death in susceptible bacteria.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100X
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives GlutaMAX
Without Additives None
Formulation Liquid, citrate buffer
Osmolality 330 to 370 mOsm/kg
pH 4.7 to 6.2
Product Line GlutaMAX
Quantity 100 mL
Regulatory Status RUO
Shipping Condition -5°C to -20°C
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.