missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Penicillin-Streptomycin (20,000 IU Pen/mL, 20,000 μg Strep/mL)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
- The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
- This solution contains 17,000 to 21,000 units/mL of penicillin and 18.0 to 23.0 mg/mL of streptomycin
Specifications
Specifications
| Content And Storage | Stored at -20°C |
| Antibiotic Spectrum | Prevent bacterial contamination of cell cultures, Gram-Positive and Gram-Negative Bacteria |
| Concentration | 20,000 IU Pen/mL, 20,000 μg Strep/mL |
| Form | Liquid |
| Product Type | Antibiotic |
| Sterility | Sterile |
| For Use With (Application) | Cell Culture |
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- Penicillin-Streptomycin Solution, 200 X
- Danger
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : streptomycin sulphate, penicillin G sodium salt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction