missing translation for 'onlineSavingsMsg'
Learn More

Melford Penicillin/Streptomycin

Product Code. 30018744
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018744 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018744 Supplier Melford Supplier No. P9356050.0
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Penicillin/Streptomycin Solution is widely used in cell culture to prevent bacterial contamination.

Penicillin/Streptomycin Solution is widely used in cell culture to prevent bacterial contamination. It is added to cell culture media to create a sterile environment for the growth of cells, inhibiting the proliferation of bacteria that may interfere with cell growth and experiments.

Commonly used in cell culture and microbiology research to prevent bacterial contamination. When added to a cell culture medium, the solution creates a sterile environment unfavorable for bacterial growth.

Specifications

CAS 3810-74-0
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.