missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals

Product Code. 15545166 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15545166 25 g 25g
15525166 1 g 1g
15535166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15545166 Supplier Thermo Scientific Chemicals Supplier No. 455690250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
  • Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis

Applications

  • Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
  • It can also be used to prevent infection in research animals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-57-8
Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Type Penicillin G Sodium Salt
pH 5.0 to 7.5
Infrared Spectrum Authentic
Specific Rotation +293° to +303° (20°C, 589nm) (c=2, H2O)
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 356.37
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Penicillin G sodium salt
compliance-icons
Product Identifier
  • Penicillin G sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.