missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Product Code. 17819436 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mU
Unit Size:
25 micro units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17819436 25 mU 25 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17819436 Supplier MP Biomedicals Supplier No. 0210054325
  • ONLINE EXCLUSIVE PRICE
    £68.10
    £63.84 / 25 micro units
    Save £4.26
    6% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Active against gram-positive and gram-negative aerobic and anaerobic bacteria. Penicillin G Potassium Salt, MP Biomedicals is a narrow spectrum antibiotic. It inhibits cell wall synthesis by binding to penicillin binding proteins (PBPs), which inhibits pe

Penicillin G is the drug of choice for Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus.

Penicillin G potassium salt is used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans. It is stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder
CAS 113-98-4
pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (literature)
Quantity 25 mU
Molecular Formula C33H36ClKN4O4S2
MDL Number MFCD00036193
Synonym Benzylpenicillin potassium salt
Solubility Information Soluble in water (100mg/mL) clear,colorless to light yellow.
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 691.34
PubChem CID 23664709
ChEBI CHEBI:7963
Formula Weight 372.48
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Penicillin G potassium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Penicillin G potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.