missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, 99%

Product Code. 10081693
Change view
Click to view available options
Quantity:
100 mU
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081693 100 mU 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10081693 Supplier Fisher BioReagents Supplier No. BP914100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 113-98-4 | C33H36ClKN4O4S2 | 691.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Penicillin G is a narrow spectrum antibiotic that is effective against gram positive bacteria
  • Useful for preventing microbial contamination in tissue culture applications
  • Functions as an antiobiotic by inhibiting cell wall synthesis 
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Penicillin-G Potassium Salt
Type Penicillin G Potassium Salt
Physical Form Solid
CAS 113-98-4
pH 5.5 to 7.5
Melting Point 214°C
Loss on Drying < 1.5%
Packaging Poly Bottle
Potency Inclusive between 1440μg/mg
Color White
Quantity 100 mU
Assay Percent Range ≥85 %
Molecular Formula C33H36ClKN4O4S2
MDL Number MFCD00036193
Merck Index 15, 7203
Synonym Benzylpenicillin potassium salt
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 691.34
PubChem CID 23664709
ChEBI CHEBI:7963
Formula Weight 372.47
Percent Purity ≥85%
Show More Show Less
compliance-icons
Product Identifier
  • Penicillin-G potassium salt
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.