Learn More
Melford Penicillin G, Potassium Salt
Description
Penicillin G potassium salt is a member of the penicillin class of antibiotics and is effective against a variety of gram-positive bacteria.
Penicillin G is effective against a wide range of Gram-positive bacteria, including Streptococcus, Staphylococcus, and some other species. However, it is not effective against Gram-negative bacteria due to their different cell wall structure.
Specifications
Specifications
| Chemical Name or Material | Penicillin G Potassium Salt |
| CAS | 113-98-4 |
| Quantity | 100 g |
| Molecular Formula | C16H17KN2O4S |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.