missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G potassium, MedChemExpress

Product Code. 16489865
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489865 250 mg 250mg
16449887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489865 Supplier Medchem Express Supplier No. HY17591

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Penicillin G potassium is a fast-acting antibiotic; used to treat bacterial infections that affect the blood, heart, lungs, joints, and genital areas.

Penicillin G potassium is a fast-acting antibiotic

Specifications

Chemical Name or Material Penicillin G potassium
Physical Form Solid
CAS 113-98-4
Color White
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C16H17KN2O4S
For Use With (Application) COVID-19-immunoregulation
Synonym Benzylpenicillin potassium
Solubility Information H2O : 100 mg/mL (268.47 mM; Need ultrasonic)
SMILES O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(CC3=CC=CC=C3)=O)N1C2=O)O[K]
Molecular Weight (g/mol) 372.48
Formula Weight 372.48
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.