missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin Antibody (Pen-9), FITC, Novus Biologicals™
Description
Penicillin Monoclonal antibody specifically detects Penicillin in All species samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Penicillin |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | Pen-9 |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin |
| Host Species | Mouse |
| Immunogen | Penicilloyl-transferrin conjugate |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?