missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin Antibody (Pen-9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14083CL1
This item is not returnable.
View return policy
Description
Penicillin Monoclonal antibody specifically detects Penicillin in Nonspecies specific samples. It is validated for ELISA
Specifications
| Penicillin | |
| Monoclonal | |
| CoraFluor 1 | |
| Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin | |
| Penicilloyl-transferrin conjugate | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| Pen-9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction