missing translation for 'onlineSavingsMsg'
Learn More

Penicillin Antibody (Pen-9), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18365082 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365082 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365082 Supplier Novus Biologicals Supplier No. NBP314083AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Penicillin Monoclonal antibody specifically detects Penicillin in All species samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Penicillin
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone Pen-9
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin
Host Species Mouse
Immunogen Penicilloyl-transferrin conjugate
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.