missing translation for 'onlineSavingsMsg'
Learn More

Penfluridol, MedChemExpress

Product Code. 16494537
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16494537

Brand: Medchem Express HYB1077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Penfluridol is a highly potent, first generation diphenylbutylpiperidine antipsychotic.

Penfluridol is a highly potent, first generation diphenylbutylpiperidine antipsychotic.

Specifications

Chemical Name or Material Penfluridol
CAS 26864-56-2
Color White
Purity Grade Notes Research
Molecular Formula C28H27ClF5NO
Quantity 50 mg
Synonym R-16341
Solubility Information DMSO : ≥ 100 mg/mL (190.85 mM)
SMILES OC1(C2=CC=C(Cl)C(C(F)(F)F)=C2)CCN(CCCC(C3=CC=C(F)C=C3)C4=CC=C(F)C=C4)CC1
Molecular Weight (g/mol) 523.97
Formula Weight 523.97
Percent Purity 98.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.