missing translation for 'onlineSavingsMsg'
Learn More

Penbutolol sulfate, MedChemExpress

Product Code. 16478678
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478678 10 mg 10mg
16474557 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478678 Supplier Medchem Express Supplier No. HYB1154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Penbutolol sulfate is able to bind to both beta-1 adrenergic receptors and beta-2 adrenergic receptors (the two subtypes), thus making it a non-selective β blocker.

Penbutolol sulfate is able to bind to both beta-1 adrenergic receptors and beta-2 adrenergic receptors (the two subtypes), thus making it a non-selective β blocker.

Specifications

Chemical Name or Material Penbutolol sulfate
CAS 38363-32-5
Color White
Purity Grade Notes Research
Molecular Formula C18H29NO2.1/2H2SO4
Quantity 10 mg
Synonym (-)-Terbuclomine
Solubility Information H2O : 5 mg/mL (14.69 mM; Need ultrasonic)
SMILES O[C@@H](CNC(C)(C)C)COC1=CC=CC=C1C2CCCC2.[0.5H2SO4]
Molecular Weight (g/mol) 340.47
Formula Weight 340.47
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.