missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PEN (mouse)
Description
PEN (mouse) is a endogenous peptide GPR83 agonist. ProSAAS-derived neuropeptide. Activates phospholipase C (PLC)-mediated signaling cascade in mouse hypothalamus.
Specifications
Specifications
| Purity | 95% |
| CAS | 1236955-25-1 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | Observed MW: 2317.62 |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?