missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598046
This item is not returnable.
View return policy
Description
PEMT Polyclonal Antibody for ELISA, ICC/IF, Western Blot
This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.
Specifications
| PEMT | |
| Polyclonal | |
| Unconjugated | |
| PEMT | |
| MGC2483; PEAMT; PEMPT; Pempt2; PEMT; PEMT2; PHOMETH; phosphatidylethanolamine N-methyltransferase; phosphatidylethanolamine N-methyltransferase-like protein; phospholipid methyltransferase; PLMT; PNMT; similar to phosphatidylethanolamine N-methyltransferase; zgc:55479 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10400 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UBM1 | |
| PEMT | |
| Recombinant Human Phosphatidylethanolamine N-methyltransferase protein (2-49AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction