missing translation for 'onlineSavingsMsg'
Learn More

Pemirolast potassium, MedChemExpress

Product Code. 16494387
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494387 50 mg 50mg
16488356 100 mg 100mg
16478518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494387 Supplier Medchem Express Supplier No. HYB0538A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.

Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.

Specifications

Chemical Name or Material Pemirolast potassium
Physical Form Solid
CAS 100299-08-9
Color Yellowish Green
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H7KN6O
For Use With (Application) COVID-19-immunoregulation
Synonym TWT-8152 BMY 26517
Solubility Information H2O : 50 mg/mL (187.76 mM; Need ultrasonic)
SMILES O=C1C(C2=N[N-]N=N2)=CN=C3N1C=CC=C3C.[K+]
Molecular Weight (g/mol) 266.3
Formula Weight 266.3
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.