missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Pembrolizumab

Product Code. 16413069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413069 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16413069 Supplier MedChemExpress Supplier No. HYP9902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pembrolizumab (MK-3475) is a humanized IgG4 antibody inhibiting the programmed cell death 1 (PD-1) receptor, used in cancer immunotherapy.

Specifications

Antigen PD-1/PD-L1
Quantity 1 mg
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Product Type Inhibitory Antibodies
Shipping Condition Shipping with dry ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.