Learn More
3M™ PELTOR™ WS™ ProTac XP Bluetooth™ Headset
Description
- Active-hearing headset with Bluetooth™, for cable free clear and reliable communications with phones or other communication devices
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- Work with both hands free while taking calls via DECT- or mobile phones
- Headset provides effective noise attenuation when sound level is high
- Supports Bluetooth™ A2DP profile for streaming high quality stereo sound
- VOX function for hands-free operation of the communication radio
Specifications
Specifications
| Product Type | Bluetooth Headset |
| Model | MT15H7P3EWS5 |
| Noise Level | 31 dB |
| Certifications/Compliance | CE Approved |