Learn More
3M™ PELTOR™ WS™ Alert™ XP Bluetooth™ Headset
Product Code. 15700058
Change view
Click to view available options
Unit Size:
Each
Model:
Helmet Mounted
Product Code. unitSize Model
15700058 Each -
15719737 Each Helmet Mounted
2 options
Product Code. 15700058
Brand: 3M™ 7100020054
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ PELTOR™ WS™ Alert™ XP Bluetooth™ Headset enables users to have wireless telephone conversations or listen music even in noisy environments.
  • Stay protected whilst taking calls wirelessly or listening to the radio - even in noisy environments
  • Level dependent function lets the user hear surrounding noise and converse with colleagues
  • Fast, easy turning of radio stations
  • Bluetooth™ for wireless hands-free calls or music streaming
  • Advanced speaker system that provides full-bodied dynamic sound
  • Battery charge via cable plugged into electrical socket or USB
  • Fast Tuning
  • Ghost voice guided interface
  • Memory/preset function
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bluetooth Headset
Includes Built-in FM Radio Receiver, Adjustable Damping Function
Noise Level 29 dB