Learn More
3M™ PELTOR™ Optime™ III Earmuffs
Description
- Ideal hearing protection against noise arising from extremely noisy environments like airports, marine engine rooms, power stations and print works
- Double casings technology helps minimize resonance effect
- Large space inside cup helps reduce moisture and heat build-up
- Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
- Easy to replace cushions and inserts helps keep them hygienically clean
Specifications
Specifications
| Color | Hi-Viz |
| Certifications/Compliance | CE Approved, EN 352-3: 2002 |
| Model | H540B-474-GB |
| Noise Reduction Rating (NRR) | 35 dB |
| Product Type | Ear Muffs |
| Dielectric | No |
| Style | Neckband |