missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ FLX2 Cable for CH-3 Listen Only Headset
Description
- For use with CH-3 Listen-Only Headsets
- Simple and reliable connection to a wide variety of handheld devices.
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | 2.5 mm mono plug |
| For Use With (Application) | Enables connection to listen-only CH-3 Headsets with Hytera, Icom, Kenwood or Motorola radios |
| Model | FLX2-203 |
| Pairs per Package | 1 Ea. |
| Product Type | FLX2 Cable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?