missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ 28 dB Earmuffs
Click to view available options
Model:
H31P3A 300
H31P3B 300
H31P3BB 300
H31P3E 300
H31P3E 300-GB
H31P3E 300-GU
H31P3K 300-GU
H31P3K-300
H31P3K-300-GU
Unit Size:
Pack of 20
Description
- Slim hearing protector ideal for use in forestry, sawmills and industry
- Earmuff cup has a recess at the top making it a more comfortable fit with Safety Helmets
- Wide, comfortable sealing rings are filled with a unique combination of fluid and foam, providing optimum sealing and ideal comfort even during long periods of use
- Earcup pivot points tilt for optimum comfort and efficiency
- Variety of helmet attachment for helmet mounting
Specifications
Specifications
| Color | Orange |
| For Use With (Application) | Forestry, sawmills, industry |
| Model | H31P3B 300 |
| Noise Reduction Rating (NRR) | 28 dB |
| Product Type | Helmet Mounted Ear Muffs |
| Dielectric | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction