missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pellino 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534465
This item is not returnable.
View return policy
Description
A suggested positive control is human liver tissue lysate. PA5-34465 can be used with blocking peptide PEP-1507.
The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.
Specifications
| Pellino 1 | |
| Polyclonal | |
| Unconjugated | |
| PELI1 | |
| 2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; E3 ubiquitin-protein ligase pellino homolog 1; Peli1; pellino 1; pellino E3 ubiquitin protein ligase 1; pellino homolog 1; pellino1; pellino-1; Pellino-related intracellular-signaling molecule; PRISM; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305549, 57162, 67245 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8C669, Q96FA3 | |
| PELI1 | |
| A 15 amino acid peptide near the carboxy terminus of human Pellino 1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction