missing translation for 'onlineSavingsMsg'
Learn More

Pellino 1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18430047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430047 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430047 Supplier Novus Biologicals Supplier No. NBP281888AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pellino 1 Polyclonal antibody specifically detects Pellino 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pellino 1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1
Host Species Rabbit
Immunogen Rabbit polyclonal Pellino 1 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human Pellino 1. The immunogen is located within amino acids 300 - 350 of Pellino 1. Amino Acid Squence: CHNWGNKEERDGKDRE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57162
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.