missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Mini holder can be autoclaved without filters installed
- The cassettes themselves cannot be autoclaved
- An optional separator plate allows processing simultaneously with up to three 0.1 m²/1.1 ft² cassettes to determine the best molecular weight cut-off in a single study on the same feed material
Specifications
Specifications
| Connectors | Feed/retentate/permeate 0.75 in. Sanitary Flange |
| Material | Stainless Steel |
| Manifold Type | Stainless Steel |
| Max. Pressure | 7 bar (100 psi) |
| Type | Silicone Gasket |
| Volume (Metric) | 5.3 to 6.4 mL |
| Length (Metric) | 14 cm |
| Width (Metric) | 11.4 cm |
| Height (Metric) | 2.6 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?