Learn More
Specac™ Pellet Die Assembly 7mm for Mini-Pellet Press
Product Code. 15279997
Encompass_Preferred
Change view
Click to view available options
Type:
Pellet Die Assembly 7mm
Product Code. Type
15279997 Pellet Die Assembly 7mm
1 options
Product Code. 15279997 Supplier Specac™ Supplier No. GS03950
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A 7mm Pellet Die and Support Ring for use within the 2 Ton Mini-Press GS03940. This die enables sample to be placed inside it to then be pressed.
Specifications
For Use With (Equipment) Mini-Pellet Press
Type Pellet Die Assembly 7mm