missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pejvakin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116276
This item is not returnable.
View return policy
Description
Pejvakin Polyclonal Antibody for Western Blot
Essential in the activity of auditory pathway neurons. Defects in PJVK are the cause of non-syndromic sensorineural deafness autosomal recessive type 59 (DFNB59). DFNB59 is a form of sensorineural hearing impairment with absent or severely abnormal auditory brainstem response but normal otoacoustic emissions (auditory neuropathy or auditory dys-synchrony). Auditory neuropathies result from a lesion in the area including the inner hair cells, connections between the inner hair cells and the cochlear branch of the auditory nerve, the auditory nerve itself and auditory pathways of the brainstem. 'Pejvak' means 'echo' in Persian.
Specifications
| Pejvakin | |
| Polyclonal | |
| Unconjugated | |
| PJVK | |
| Autosomal recessive deafness type 59 protein; autosomal recessive deafness type 59 protein homolog; deafness, autosomal recessive 59; deafness, autosomal recessive 59 (human); Dfnb59; DFNB59 deafness; Gm1001; Pejvakin; Pjvk | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 494513 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q0ZLH3 | |
| PJVK | |
| KLH conjugated synthetic peptide between 170-197 amino acids from the Central region of human PJVK. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction