missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Spacious peg design allows easy viewing of tube contents
- Use as a drying rack for glass plates or upside-down tubes
- Pegs remain straight even after repeated autoclaving
- Embossed alphanumeric identification for quick location and retrieval of tubes
- Unique tear drop well design allows for better retention of tubes
- Autoclavable
Specifications
Specifications
| Description | Peg Tube Rack |
| Autoclavable | Autoclavable |
| Color | White |
| Holds | Tubes |
| Material | Fiberglass Reinforced |
| Shape | Rectangular |
| Stackable | No |
| Sterility | Non-sterile |
| For Use With (Application) | Tubes |
| Model | HS24312A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?