missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEG (methoxy group) Polyclonal Antibody
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
PEG recruits the serine/threonine-protein phosphatase PP1 to dephosphorylate the translation initiation factor eIF-2A/EIF2S1, thereby reversing the shut-off of protein synthesis initiated by stress-inducible kinases and facilitating recovery of cells from stress. Down-regulates the TGF-beta signaling pathway by promoting dephosphorylation of TGFB1 by PP1. May promote apoptosis by inducing TP53 phosphorylation on 'Ser-15'.
Specifications
Specifications
| Antigen | PEG (methoxy group) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene Alias | Poly Ethylene Glycol; Polyethylene Glycol |
| Host Species | Rabbit |
| Immunogen | The immunogen used to generate this antibody corresponds to PEG (methoxy group). |
| Purification Method | Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?