missing translation for 'onlineSavingsMsg'
Learn More

Peforelin Trifluoroacetic Acid Salt(>90%), TRC

Product Code. 30420007
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420007 5 mg 5mg
30414911 1 mg 1mg
30412554 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420007 Supplier Toronto Research Chemicals Supplier No. TRCP2183355MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Peforelin Trifluoroacetic Acid Salt
Molecular Formula C59H74N18O14 .C2HF3O2
Quantity 5 mg
Synonym 3-L-Tryptophan-5-L-histidine-6-L-aspartic Acid Luteinizing Hormone-releasing Factor I (Petromyzon Marinus) Trifluoroacetic Acid Salt, Luteinizing Hormone-releasing Factor III (Petromyzon Marinus) Trifluoroacetic Acid Salt, 5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-L-histidyl-L-α-aspartyl-L-tryptophyl-L-lysyl-L-prolylglycinamide Trifluoroacetic Acid Salt, Lamprey GnRH III Trifluoroacetic Acid Salt, Lamprey LH-RH III Trifluoroacetic Acid Salt
InChI Formula InChI=1S/C59H74N18O14.C2HF3O2/c60-16-6-5-12-40(59(91)77-17-7-13-47(77)58(90)66-27-48(61)79)70-52(84)41(18-31-23-64-37-10-3-1-8-35(31)37)72-56(88)45(22-50(81)82)75-55(87)44(21-34-26-63-30-68-34)74-57(89)46(28-78)76-53(85)42(19-32-24-65-38-11-4-2-9-36(32)38)71-54(86)43(20-33-25-62-29-67-33)73-51(83)39-14-15-49(80)69-39;3-2(4,5)1(6)7/h1-4,8-11,23-26,29-30,39-47,64-65,78H,5-7,12-22,27-28,60H2,(H2,61,79)(H,62,67)(H,63,68)(H,66,90)(H,69,80)(H,70,84)(H,71,86)(H,72,88)(H,73,83)(H,74,89)(H,75,87)(H,76,85)(H,81,82);(H,6,7)/t39-,40-,41-,42-,43-,44-,45-,46-,47-;/m0./s1
SMILES O=C(O)C(F)(F)F.O=C([C@@H](NC([C@H](CC1=CN=CN1)NC([C@@H](NC([C@H](CC2=CNC3=CC=CC=C32)NC([C@H](CC4=CN=CN4)NC([C@H]5NC(CC5)=O)=O)=O)=O)CO)=O)=O)CC(O)=O)N[C@H](C(N[C@H](C(N6CCC[C@H]6C(NCC(N)=O)=O)=O)CCCCN)=O)CC7=CNC8=CC=CC=C87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.