missing translation for 'onlineSavingsMsg'
Learn More

Pefloxacin Mesylate, TRC

Product Code. 30416170
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30416170

Brand: Toronto Research Chemicals TRCP21830050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pefloxacin mesilate
CAS 70458-95-6
Molecular Formula C17 H20 F N3 O3 . C H4 O3 S
Quantity 50 mg
Synonym 3-Quinolinecarboxylic acid','1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-','methanesulfonate (1:1) (ACI), 3-Quinolinecarboxylic acid','1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-','monomethanesulfonate (9CI), 41-982RP, Peflacin, Pefloxacin mesylate, Pefloxacin methanesulfonate, 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid methanesulfonate (1:1), 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid monomethanesulfonate, Pefloxacin mesilate
InChI Formula InChI=1S/C17H20FN3O3.CH4O3S/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4)
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3.CS(=O)(=O)O
IUPAC Name 1-ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid
Molecular Weight (g/mol) 429.46
Formula Weight 429.137
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.