missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pefloxacin Mesylate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Pefloxacin mesilate |
| CAS | 70458-95-6 |
| Molecular Formula | C17 H20 F N3 O3 . C H4 O3 S |
| Quantity | 250 mg |
| Synonym | 3-Quinolinecarboxylic acid','1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-','methanesulfonate (1:1) (ACI), 3-Quinolinecarboxylic acid','1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-','monomethanesulfonate (9CI), 41-982RP, Peflacin, Pefloxacin mesylate, Pefloxacin methanesulfonate, 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid methanesulfonate (1:1), 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid monomethanesulfonate, Pefloxacin mesilate |
| InChI Formula | InChI=1S/C17H20FN3O3.CH4O3S/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4) |
| SMILES | CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3.CS(=O)(=O)O |
| IUPAC Name | 1-ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid |
| Molecular Weight (g/mol) | 429.46 |
| Formula Weight | 429.137 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction