missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pefloxacin mesylate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Pefloxacin mesylate is a an antibacterial agent and prevents bacterial DNA replication by inhibiting DNA gyrase (topoisomerse)
Specifications
Specifications
| Chemical Name or Material | Pefloxacin mesylate |
| Physical Form | Solid |
| CAS | 70458-95-6 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H24FN3O6S |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | Pefloxacinium mesylate |
| Solubility Information | H2O : ≥ 100 mg/mL (232.85 mM) ∣DMSO : 12.5 mg/mL (29.11 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction