missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584508
This item is not returnable.
View return policy
Description
Immunogen sequence: PPGSYYPGPP NSGGQYGSGL PPGGGYGGPA PGGPYGPPAG GGPYGHPNPG MFPSGTPGGP YGGAA Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 69%.
PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
Specifications
| PEF1 | |
| Polyclonal | |
| Unconjugated | |
| PEF1 | |
| 2600002E23Rik; ABP32; PEF protein with a long N-terminal hydrophobic domain; Pef1; PEF1A; Peflin; peflin; LOW QUALITY PROTEIN: peflin; penta-EF hand domain containing 1; penta-EF hand domain-containing protein 1; penta-EF-hand domain containing 1; UNQ1845/PRO3573; zgc:100787 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 553115 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBV8 | |
| PEF1 | |
| Recombinant protein corresponding to Human PEF1. Recombinant protein control fragment (Product #RP-105196). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction